siRNA Details
Gene Symbol: Phyh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053674.1 | NP_446126.1 | 4 | 359 | ||
GenBank mRNA | AF121345.1 | - | 4 | 359 | ||
BC086573.1 | - | 4 | 354 | 7306754 | ||
FQ216943.1 | - | 2 | 191 | |||
FQ224924.1 | - | 2 | 191 |
