siRNA Details
Gene Symbol: Insl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053680.1 | NP_446132.1 | 2 | 480 | ||
XM_006252820.1 | XP_006252882.1 | 3 | 1136 | |||
GenBank mRNA | AF139918.2 | - | 2 | 480 | ||
BC098653.1 | - | 2 | 1323 | 7386551 |

More Information
Other Species Targeted

Gene Ontology Categories:
