siRNA Details
Gene Symbol: Dmrt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053706.1 | NP_446158.1 | 4 | 1016 | ||
XM_008760301.1 | XP_008758523.1 | 4 | 1106 | |||
GenBank mRNA | AF379608.1 | - | 4 | 1016 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
