siRNA Details
Gene Symbol: Hdgf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053707.2 | NP_446159.1 | 4 | 674 | ||
GenBank mRNA | AF389348.1 | - | 4 | 403 | ||
AF448810.1 | - | 4 | 403 | |||
BC070943.1 | - | 4 | 674 | 7103046 | ||
FQ231256.1 | - | 5 | 437 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
