siRNA Details
Gene Symbol: Cntn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053746.1 | NP_446198.1 | 6, 7 | 893 | ||
XM_008765902.1 | XP_008764124.1 | 6, 7 | 958 | |||
GenBank mRNA | D87212.1 | - | 6, 7 | 893 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
