siRNA Details
Gene Symbol: Aurkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053749.1 | NP_446201.1 | 4 | 352 | ||
XM_006246569.2 | XP_006246631.1 | 4 | 370 | |||
XM_006246570.2 | XP_006246632.1 | 3 | 257 | |||
XM_008767757.1 | XP_008765979.1 | 4 | 267 | |||
XM_008767758.1 | XP_008765980.1 | 4 | 213 | |||
GenBank mRNA | BC097297.1 | - | 4 | 270 | 7454285 | |
D89731.1 | - | 4 | 352 |
