siRNA Details
Gene Symbol: Wap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053751.4 | NP_446203.1 | 3 | 315 | ||
XM_008770244.1 | XP_008768466.1 | 3 | 682 | |||
GenBank mRNA | J00801.1 | - | 3 | 312 | ||
J00802.1 | - | 3 | 315 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
