siRNA Details
Gene Symbol: Suclg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053752.2 | NP_446204.2 | 7 | 776 | ||
XM_008762973.1 | XP_008761195.1 | 6 | 689 | |||
GenBank mRNA | BC061537.1 | - | 7 | 780 | 6920009 | |
FQ212668.1 | - | 7 | 773 | |||
FQ214532.1 | - | 7 | 735 | |||
FQ214959.1 | - | 7 | 765 | |||
FQ215028.1 | - | 7 | 734 | |||
FQ215469.1 | - | 7 | 773 | |||
FQ216893.1 | - | 7 | 766 | |||
FQ217244.1 | - | 7 | 790 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
