siRNA Details
Gene Symbol: Suclg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053752.2 | NP_446204.2 | 8 | 872 | ||
XM_008762973.1 | XP_008761195.1 | 7 | 785 | |||
GenBank mRNA | BC061537.1 | - | 8 | 876 | 6920009 | |
FQ212668.1 | - | 8 | 869 | |||
FQ214532.1 | - | 8 | 831 | |||
FQ214959.1 | - | 8 | 861 | |||
FQ215028.1 | - | 8 | 830 | |||
FQ215469.1 | - | 8 | 869 | |||
FQ216893.1 | - | 8 | 862 | |||
FQ217244.1 | - | 8 | 886 |

More Information
Other Species Targeted

Gene Ontology Categories:
