siRNA Details
Gene Symbol: Slc26a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053755.2 | NP_446207.1 | 18 | 2199 | ||
GenBank mRNA | AF337809.1 | - | 18 | 2204 |

More Information
Other Species Targeted

Gene Ontology Categories:
