siRNA Details
Gene Symbol: Six1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053759.1 | NP_446211.1 | 1, 2 | 1178 | ||
GenBank mRNA | AF323957.1 | - | 1, 2 | 545 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009189, AK131925, X80339, AK144753, BC023304, and others.
Gene Ontology Categories:
