siRNA Details
Gene Symbol: Hmgn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025624.2 | NP_001020795.1 | 5 | 341 | ||
GenBank mRNA | BC099178.1 | - | 5 | 307 | 7383003 | |
BC099815.1 | - | 5 | 316 | 7929654 | ||
CB614313.1 | - | 5 | 350 | |||
FQ213300.1 | - | 5 | 309 | |||
FQ213727.1 | - | 5 | 300 | |||
FQ214234.1 | - | 3 | 136 | |||
FQ214394.1 | - | 5 | 300 | |||
FQ214422.1 | - | 5 | 311 | |||
FQ215102.1 | - | 5 | 317 | |||
FQ216264.1 | - | 5 | 335 | |||
FQ216489.1 | - | 5 | 272 | |||
FQ217912.1 | - | 5 | 311 | |||
FQ219745.1 | - | 5 | 376 | |||
FQ219836.1 | - | 5 | 338 | |||
FQ220601.1 | - | 5 | 352 | |||
FQ221300.1 | - | 5 | 282 | |||
FQ223249.1 | - | 5 | 300 | |||
FQ229179.1 | - | 5 | 300 | |||
FQ229184.1 | - | 5 | 274 |
Gene Symbol: LOC100360316 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751587.3 | XP_003751635.1 | 1 | 307 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
