siRNA Details
Gene Symbol: LOC100360316 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751587.3 | XP_003751635.1 | 1 | 1152 |
Gene Symbol: Hmgn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025624.2 | NP_001020795.1 | 6 | 1189 | ||
GenBank mRNA | BC099178.1 | - | 6 | 1156 | 7383003 | |
BC099815.1 | - | 6 | 1164 | 7929654 | ||
FQ213300.1 | - | 6 | 1156 | |||
FQ213727.1 | - | 6 | 1149 | |||
FQ214234.1 | - | 4 | 982 | |||
FQ214394.1 | - | 6 | 1148 | |||
FQ214422.1 | - | 6 | 1159 | |||
FQ215102.1 | - | 6 | 1166 | |||
FQ216264.1 | - | 6 | 1182 | |||
FQ216489.1 | - | 6 | 1121 | |||
FQ217912.1 | - | 6 | 1158 | |||
FQ219745.1 | - | 6 | 1224 | |||
FQ219836.1 | - | 6 | 1186 | |||
FQ220601.1 | - | 6 | 1200 | |||
FQ221300.1 | - | 6 | 1130 | |||
FQ223249.1 | - | 6 | 1147 | |||
FQ229179.1 | - | 6 | 1148 | |||
FQ229184.1 | - | 6 | 1122 |

More Information
Other Species Targeted

Gene Ontology Categories:
