siRNA Details
Gene Symbol: Hps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_040669.1 | NP_414541.1 | 18, 19 | 2029 | ||
XM_006231378.2 | XP_006231440.1 | 19, 20 | 2156 | |||
XM_006231379.2 | XP_006231441.1 | 19, 20 | 2282 | |||
XM_006231380.2 | XP_006231442.1 | 19, 20 | 2112 | |||
XM_006231381.2 | XP_006231443.1 | 19, 20 | 2223 | |||
GenBank mRNA | AF333325.1 | - | 18, 19 | 2029 |
