siRNA Details
Gene Symbol: Cyp27b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053763.1 | NP_446215.1 | 1 | 110 | ||
XM_008765374.1 | XP_008763596.1 | 2 | 1982 | |||
GenBank mRNA | AB001992.1 | - | 1 | 110 | ||
AF000139.1 | - | 1 | 102 |
Gene Symbol: LOC100909462 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773774.1 | XP_008771996.1 | 870 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
