siRNA Details
Gene Symbol: Mapre1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138509.3 | NP_612518.2 | 3 | 280 | ||
XM_006235294.2 | XP_006235356.1 | 3 | 263 | |||
XM_006235295.2 | XP_006235357.1 | 3 | 333 | |||
GenBank mRNA | BC081726.1 | - | 3 | 280 | 7192469 |
