siRNA Details
Gene Symbol: Uox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053768.2 | NP_446220.1 | 6 | 705 | ||
XM_006233450.2 | XP_006233512.1 | 7 | 896 | |||
GenBank mRNA | BC088112.1 | - | 6 | 707 | 7325917 | |
FQ209540.1 | - | 6 | 718 | |||
FQ209683.1 | - | 6 | 721 | |||
FQ210147.1 | - | 5 | 627 | |||
FQ210270.1 | - | 6 | 719 | |||
FQ218233.1 | - | 6 | 721 | |||
FQ218518.1 | - | 6 | 719 | |||
FQ218662.1 | - | 5 | 447 | |||
FQ218801.1 | - | 6 | 719 | |||
FQ218875.1 | - | 6 | 720 | |||
FQ219119.1 | - | 6 | 719 | |||
FQ219150.1 | - | 6 | 719 | |||
FQ219398.1 | - | 6 | 721 | |||
J03959.1 | - | 6 | 663 | |||
M24396.1 | - | 6 | 706 | |||
X07497.1 | - | 5 | 649 | |||
X13098.1 | - | 6 | 706 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
