siRNA Details
Gene Symbol: Shc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105743.1 | NP_001099213.1 | 4, 5 | 730 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009167, BC103612, BC105644, BC103611, U46854, and others.
Gene Ontology Categories:
