siRNA Details
Gene Symbol: Eif2ak4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105744.2 | NP_001099214.2 | 33 | 4572 | ||
GenBank mRNA | CF108241.1 | - | 3 | 321 | ||
FQ228781.1 | - | 6 | 707 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
