siRNA Details
Gene Symbol: Eif2ak4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105744.2 | NP_001099214.2 | 38 | 4961 | ||
GenBank mRNA | CF108241.1 | - | 8 | 710 | ||
FQ228781.1 | - | 11 | 1096 |

More Information
Other Species Targeted

Gene Ontology Categories:
