siRNA Details
Gene Symbol: Sorbs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053770.1 | NP_446222.1 | 22 | 4189 | ||
GenBank mRNA | AF026505.1 | - | 22 | 4189 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sorbs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053770.1 | NP_446222.1 | 22 | 4189 | ||
GenBank mRNA | AF026505.1 | - | 22 | 4189 |