siRNA Details
Gene Symbol: Zfp37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_058209.2 | NP_478116.2 | 1324 | |||
GenBank mRNA | AF072439.1 | - | 1355 | |||
BC166844.1 | - | 1298 |
Gene Symbol: LOC100363405 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763746.1 | XP_008761968.1 | 1305 |

More Information
Other Species Targeted

Gene Ontology Categories:
