siRNA Details
Gene Symbol: Usp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053774.2 | NP_446226.2 | 10 | 1619 | ||
XM_006242861.2 | XP_006242923.1 | 10 | 1780 | |||
XM_006242863.2 | XP_006242925.1 | 9 | 1071 | |||
XM_008766138.1 | XP_008764360.1 | 9 | 920 | |||
XM_008766139.1 | XP_008764361.1 | 9 | 923 | |||
GenBank mRNA | AF106658.1 | - | 9 | 1047 | ||
AF106659.1 | - | 10 | 1619 | |||
AF202453.1 | - | 9 | 1492 | |||
AF202454.1 | - | 9 | 826 | |||
BC085719.1 | - | 10 | 1593 | 7193298 |
