siRNA Details
Gene Symbol: Mapk8ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053777.1 | NP_446229.1 | 5 | 1390 | ||
GenBank mRNA | AF092450.1 | - | 4 | 1125 | ||
AF108959.1 | - | 4 | 1328 | |||
AF109772.1 | - | 4 | 1237 | |||
AF109773.1 | - | 4 | 1300 | |||
AF109774.1 | - | 5 | 1390 | |||
DQ377223.1 | - | 4 | 1200 |

More Information
Other Species Targeted

Gene Ontology Categories:
