siRNA Details
Gene Symbol: Serpini1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053779.1 | NP_446231.1 | 3, 4 | 587 | ||
XM_008761076.1 | XP_008759298.1 | 4, 5 | 874 | |||
GenBank mRNA | AF193014.1 | - | 3, 4 | 587 | ||
AF193015.1 | - | 3, 4 | 593 | |||
BC061536.1 | - | 3, 4 | 601 | 6920827 |
