siRNA Details
Gene Symbol: Dars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053799.2 | NP_446251.1 | 7, 8 | 560 | ||
XM_006249684.2 | XP_006249746.1 | 7, 8 | 636 | |||
GenBank mRNA | BC072534.1 | - | 7, 8 | 685 | 7098427 | |
J04487.1 | - | 7, 8 | 561 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
