siRNA Details
Gene Symbol: Fgf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053809.1 | NP_446261.1 | 3 | 431 | ||
XM_006230640.2 | XP_006230702.1 | 3 | 561 | |||
GenBank mRNA | AB079673.1 | - | 3 | 431 | ||
AF260830.1 | - | 2 | 97 |

More Information
Other Species Targeted

Gene Ontology Categories:
