siRNA Details
Gene Symbol: S100a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053822.2 | NP_446274.2 | 1, 2 | 169 | ||
XM_006232565.2 | XP_006232627.1 | 2, 3 | 269 | |||
GenBank mRNA | L18891.1 | - | 1, 2 | 175 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
