siRNA Details
Gene Symbol: Foxj1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053832.1 | NP_446284.1 | 583 | |||
GenBank mRNA | L36388.1 | - | 583 |

More Information
Other Species Targeted

Gene Ontology Categories:
