siRNA Details
Gene Symbol: Fcgr2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053843.2 | NP_446295.2 | 5 | 646 | ||
XM_006250303.2 | XP_006250365.1 | 5 | 699 | |||
XM_006250305.2 | XP_006250367.1 | 5 | 798 | |||
GenBank mRNA | BC060534.1 | - | 423 | 6886727 | ||
L08446.1 | - | 5 | 646 |
Gene Symbol: LOC103693683 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769773.1 | XP_008767995.1 | 4 | 451 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
