siRNA Details
Gene Symbol: Map3k8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053847.1 | NP_446299.1 | 2 | 592 | ||
XM_006254048.2 | XP_006254110.1 | 2 | 612 | |||
GenBank mRNA | FQ217924.1 | - | 2 | 625 | ||
M94454.1 | - | 2 | 592 |
