siRNA Details
Gene Symbol: Opcml | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053848.1 | NP_446300.1 | 4 | 1056 | ||
XM_008766041.1 | XP_008764263.1 | 2 | 214 | |||
GenBank mRNA | M88709.1 | - | 1206 | |||
M88710.1 | - | 4 | 887 | |||
M88711.1 | - | 4 | 1056 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
