siRNA Details
Gene Symbol: Opcml | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053848.1 | NP_446300.1 | 4, 5 | 1137 | ||
XM_008766041.1 | XP_008764263.1 | 2, 3 | 295 | |||
GenBank mRNA | M88709.1 | - | 1287 | |||
M88710.1 | - | 4, 5 | 968 | |||
M88711.1 | - | 4, 5 | 1137 |

More Information
Other Species Targeted

Gene Ontology Categories:
