siRNA Details
Gene Symbol: Nat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053854.1 | NP_446306.1 | 3 | 479 | ||
XM_006253007.1 | XP_006253069.1 | 2 | 2070 | |||
XM_006253008.1 | XP_006253070.1 | 1 | 165 | |||
XM_006253009.1 | XP_006253071.1 | 2 | 1257 | |||
XM_008771063.1 | XP_008769285.1 | 2 | 244 | |||
GenBank mRNA | U01348.1 | - | 3 | 479 | ||
U17261.1 | - | 1 | 174 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
