siRNA Details
Gene Symbol: Ccl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053858.1 | NP_446310.1 | 2, 3 | 193 | ||
XM_006246966.2 | XP_006247028.1 | 2, 3 | 317 | |||
GenBank mRNA | EF121996.1 | - | 2, 3 | 193 | ||
EF121997.1 | - | 2, 3 | 193 | |||
U06434.1 | - | 2, 3 | 193 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
