siRNA Details
Gene Symbol: Vcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053864.2 | NP_446316.1 | 3 | 471 | ||
GenBank mRNA | BC060518.1 | - | 3 | 471 | 5597843 | |
U11760.1 | - | 3 | 503 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009503, AK214159, AK159509, AK028264, AK169140, and others.
Gene Ontology Categories:
