siRNA Details
Gene Symbol: Vcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053864.2 | NP_446316.1 | 2 | 279 | ||
GenBank mRNA | BC060518.1 | - | 2 | 279 | 5597843 | |
U11760.1 | - | 2 | 311 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
