siRNA Details
Gene Symbol: Tpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053867.1 | NP_446319.1 | 3 | 336 | ||
GenBank mRNA | BC086358.1 | - | 3 | 347 | 7303675 | |
FQ210153.1 | - | 3 | 364 | |||
FQ210845.1 | - | 2 | 250 | |||
FQ217312.1 | - | 3 | 364 | |||
FQ217485.1 | - | 3 | 364 | |||
FQ220913.1 | - | 3 | 367 | |||
FQ221184.1 | - | 3 | 364 | |||
FQ221191.1 | - | 3 | 353 | |||
FQ221220.1 | - | 3 | 364 | |||
FQ221227.1 | - | 3 | 364 | |||
FQ221357.1 | - | 3 | 364 | |||
FQ221503.1 | - | 3 | 364 | |||
FQ221524.1 | - | 3 | 364 | |||
FQ221695.1 | - | 3 | 364 | |||
FQ221696.1 | - | 3 | 365 | |||
FQ222296.1 | - | 3 | 364 | |||
FQ222704.1 | - | 3 | 363 | |||
FQ222827.1 | - | 3 | 364 | |||
FQ223172.1 | - | 3 | 365 | |||
FQ223294.1 | - | 3 | 371 | |||
FQ223612.1 | - | 3 | 353 | |||
FQ224095.1 | - | 3 | 364 | |||
FQ224271.1 | - | 3 | 365 | |||
FQ224320.1 | - | 3 | 363 | |||
FQ224622.1 | - | 3 | 300 | |||
FQ224742.1 | - | 3 | 364 | |||
FQ226252.1 | - | 3 | 364 | |||
FQ227715.1 | - | 3 | 365 | |||
FQ227806.1 | - | 3 | 393 | |||
FQ228514.1 | - | 3 | 364 | |||
FQ228556.1 | - | 3 | 364 | |||
FQ228624.1 | - | 3 | 364 | |||
FQ228630.1 | - | 3 | 364 | |||
FQ228644.1 | - | 3 | 361 | |||
FQ229028.1 | - | 3 | 364 | |||
FQ229093.1 | - | 3 | 364 | |||
FQ229217.1 | - | 3 | 364 | |||
FQ229279.1 | - | 3 | 364 | |||
FQ229346.1 | - | 3 | 364 | |||
FQ229353.1 | - | 3 | 364 | |||
FQ230785.1 | - | 3 | 364 | |||
FQ231339.1 | - | 3 | 362 | |||
FQ231778.1 | - | 3 | 364 | |||
FQ232593.1 | - | 3 | 364 | |||
FQ232768.1 | - | 3 | 365 | |||
FQ233320.1 | - | 3 | 395 | |||
FQ233909.1 | - | 3 | 367 | |||
FQ234047.1 | - | 3 | 354 | |||
FQ234186.1 | - | 3 | 365 | |||
FQ234298.1 | - | 3 | 364 | |||
FQ234781.1 | - | 3 | 364 | |||
FQ234990.1 | - | 3 | 364 | |||
FQ235307.1 | - | 3 | 364 | |||
U20525.1 | - | 3 | 336 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
