siRNA Details
Gene Symbol: Cap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053874.1 | NP_446326.1 | 8 | 837 | ||
XM_006253753.2 | XP_006253815.1 | 9 | 946 | |||
XM_006253754.2 | XP_006253816.1 | 9 | 1059 | |||
GenBank mRNA | U31935.1 | - | 8 | 837 |
