siRNA Details
Gene Symbol: Hnrnpm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109911.1 | NP_001103381.1 | 11 | 1039 | ||
NM_053876.2 | NP_446328.2 | 12 | 922 | |||
XM_006241116.2 | XP_006241178.1 | 11 | 2229 | |||
XM_008765198.1 | XP_008763420.1 | 9 | 769 | |||
GenBank mRNA | AY724543.1 | - | 11 | 1026 | ||
BC088317.1 | - | 7 | 550 | 7325202 | ||
U32577.2 | - | 12 | 922 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_029804, NM_001109913, AK168347, AK133911, AK169101, and others.
Gene Ontology Categories:
