siRNA Details
Gene Symbol: Dusp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053883.2 | NP_446335.1 | 2 | 1195 | ||
GenBank mRNA | AY724528.1 | - | 2 | 1196 | ||
BC087003.1 | - | 2 | 1195 | 7124105 | ||
U42627.1 | - | 2 | 1122 | |||
X94185.1 | - | 2 | 988 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
