siRNA Details
Gene Symbol: Atp6v1f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053884.1 | NP_446336.1 | 1, 2 | 211 | ||
GenBank mRNA | U43175.1 | - | 1, 2 | 211 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Atp6v1f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053884.1 | NP_446336.1 | 1, 2 | 211 | ||
GenBank mRNA | U43175.1 | - | 1, 2 | 211 |