siRNA Details
Gene Symbol: Myt1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053888.1 | NP_446340.1 | 24 | 4247 | ||
GenBank mRNA | U48809.1 | - | 24 | 4267 | ||
U67081.1 | - | 2349 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001093775, NM_001093776, NM_001093778, NM_008666, BC094438, and others.
Gene Ontology Categories:
