siRNA Details
Gene Symbol: Ppp1r12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053890.1 | NP_446342.1 | 20 | 2620 | ||
XM_006241308.1 | XP_006241370.1 | 22 | 3063 | |||
XM_006241309.1 | XP_006241371.1 | 22 | 3063 | |||
XM_006241311.1 | XP_006241373.1 | 21 | 2949 | |||
XM_006241312.1 | XP_006241374.1 | 21 | 2895 | |||
XM_006241313.1 | XP_006241375.1 | 21 | 2883 | |||
XM_006241314.1 | XP_006241376.1 | 21 | 2949 | |||
XM_006241315.1 | XP_006241377.1 | 20 | 2769 | |||
XM_006241316.2 | XP_006241378.1 | 20 | 3095 | |||
GenBank mRNA | S74907.1 | - | 20 | 2620 |
