siRNA Details
Gene Symbol: Dnase1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053907.1 | NP_446359.1 | 8 | 1088 | ||
GenBank mRNA | AF039852.1 | - | 8 | 1038 | ||
BC088122.1 | - | 8 | 1086 | 7325326 | ||
FQ226037.1 | - | 8 | 915 | |||
FQ227612.1 | - | 8 | 915 | |||
FQ230283.1 | - | 8 | 915 | |||
FQ230990.1 | - | 8 | 915 | |||
FQ231646.1 | - | 8 | 914 | |||
FQ231744.1 | - | 8 | 914 | |||
FQ231870.1 | - | 8 | 918 | |||
FQ231939.1 | - | 8 | 915 | |||
FQ231976.1 | - | 8 | 971 | |||
FQ233228.1 | - | 8 | 971 | |||
FQ234709.1 | - | 8 | 972 | |||
U75689.1 | - | 8 | 1088 |
Gene Symbol: LOC103690049 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770646.1 | XP_008768868.1 | 6 | 823 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
