siRNA Details
Gene Symbol: Cyth1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053910.1 | NP_446362.1 | 3 | 212 | ||
XM_006247784.2 | XP_006247846.1 | 3 | 256 | |||
XM_006247785.2 | XP_006247847.1 | 3 | 1347 | |||
XM_008768306.1 | XP_008766528.1 | 2 | 2652 | |||
XM_008768307.1 | XP_008766529.1 | 2 | 209 | |||
GenBank mRNA | BC166417.1 | - | 3 | 212 | 9028626 | |
U83895.1 | - | 3 | 212 |
