siRNA Details
Gene Symbol: Cga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053918.2 | NP_446370.2 | 2, 3 | 194 | ||
GenBank mRNA | BC063160.1 | - | 2, 3 | 198 | 6922315 | |
D00575.1 | - | 2, 3 | 168 | |||
J00757.1 | - | 1, 2 | 153 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
