siRNA Details
Gene Symbol: Slc7a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053929.1 | NP_446381.1 | 6, 7 | 780 | ||
XM_006228875.2 | XP_006228937.1 | 7, 8 | 882 | |||
XM_006228876.2 | XP_006228938.1 | 7, 8 | 1262 | |||
XM_006228877.2 | XP_006228939.1 | 6, 7 | 1141 | |||
XM_006228878.2 | XP_006228940.1 | 3, 4 | 447 | |||
XM_008759103.1 | XP_008757325.1 | 6, 7 | 1166 | |||
XM_008759104.1 | XP_008757326.1 | 6, 7 | 821 | |||
GenBank mRNA | AB029559.1 | - | 6, 7 | 780 | ||
BC081750.1 | - | 5, 6 | 1196 | |||
BC098909.1 | - | 6, 7 | 770 | 7109606 |
