siRNA Details
Gene Symbol: Lpar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053936.3 | NP_446388.1 | 3 | 222 | ||
XM_006238195.2 | XP_006238257.1 | 4 | 488 | |||
XM_006238197.2 | XP_006238259.1 | 3 | 2487 | |||
XM_006238200.2 | XP_006238262.1 | 3 | 1969 | |||
XM_006238201.2 | XP_006238263.1 | 3 | 404 | |||
XM_008763732.1 | XP_008761954.1 | 5 | 560 | |||
GenBank mRNA | AF014418.1 | - | 3 | 220 | ||
AF090347.1 | - | 3 | 240 | |||
BC089227.1 | - | 3 | 222 | 7320884 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
