siRNA Details
Gene Symbol: Pcdha8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053942.1 | NP_446394.1 | 1 | 531 | ||
GenBank mRNA | AB113391.1 | - | 1 | 531 | ||
AF177688.1 | - | 1 | 145 | |||
AY573982.1 | - | 1 | 531 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
